Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CCL2/JE/MCP-1 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB4791G100UG
This item is not returnable.
View return policy
Description
CCL2/JE/MCP-1 Monoclonal specifically detects CCL2/JE/MCP-1 in Mouse samples. It is validated for Flow Cytometry.Specifications
CCL2/JE/MCP-1 | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
C-C motif chemokine ligand 2, CCL2, GDCF-2, HC11, HSMCR30, MCAF, Mcp1, MCP-1, SCYA2, SMC-CF | |
Rat | |
100 μg | |
Primary | |
Detects mouse CCL2 in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Flow Cytometry | |
123616R | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P10148 | |
CCL2 | |
E. coli-derived recombinant mouse CCL2 Gln24-Arg96 Accession # P10148 | |
RUO | |
6347 | |
Mouse | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction