Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CCL2/JE/MCP-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB479500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB479500 500 μg
MAB479SP 25 μg
MAB479100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB479500 Supplier R&D Systems Supplier No. MAB479500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 16 publications

CCL2/JE/MCP-1 Monoclonal specifically detects CCL2/JE/MCP-1 in Mouse samples. It is validated for Neutralization.

Specifications

Antigen CCL2/JE/MCP-1
Applications Neutralization
Classification Monoclonal
Clone 123616
Conjugate Unconjugated
Dilution Neutralization 0.75-3.0 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P10148
Gene Alias C-C motif chemokine ligand 2, CCL2, GDCF-2, HC11, HSMCR30, MCAF, Mcp1, MCP-1, SCYA2, SMC-CF
Gene Symbols Ccl2
Host Species Rat
Immunogen E. coli-derived recombinant mouse CCL2/JE/MCP-1 Gln24-Arg96 Accession # P10148
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6347
Test Specificity Detects mouse CCL2/JE/MCP-1 in ELISAs. Does not cross-react with recombinant human (rh) CCL1, 2, 3, 4, 5, 7, 8, 11, 13, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, 26, 27, 28, rmCCL1, 3, 4, 5, 6, 7, 9/10/MIP-1 gamma , 11, 12, 17, 19, 20, 21, 22, 24, 25, 27, 28, or rrCCL20.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.