Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CCL20/MIP-3 alpha Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications
$214.00 - $2020.00
Specifications
| Antigen | CCL20/MIP-3 alpha |
|---|---|
| Clone | 114906 |
| Dilution | Western Blot 2 ug/mL |
| Applications | Western Blot, Neutralization |
| Classification | Monoclonal |
Description
CCL20/MIP-3 alpha Monoclonal specifically detects CCL20/MIP-3 alpha in Mouse samples. It is validated for Western Blot.Specifications
| CCL20/MIP-3 alpha | |
| Western Blot 2 ug/mL | |
| Monoclonal | |
| Rat | |
| Mouse | |
| O89093 | |
| 6364 | |
| E. coli-derived recombinant mouse CCL20/MIP-3 alpha Ala28-Met97 Accession # O89093 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse CCL20/MIP-3 alpha in direct ELISAs and Western blots. In direct ELISAs, 12% cross-reactivity with recombinant rat (rr) CCL20, 6% cross-reactivity with recombinant mouse (rm) CCL12 and recombinant human (rh) CCL25, and 3% cross-reactivity with with rmCCL9/10/MIP-1 gamma is observed. No cross-reactivity with recombinant cotton rat CCL3, 4, 5, rhCCL1, 2, 3, 4, 5, 7, 8, 1113, 14, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 26, 27, 28, rmCCL1, 2, 3, 4, 5, 6, 7, 8, 11, 17, 19, 21, 22, 24, 25, 27, or 28 is observed. |
| 114906 | |
| Western Blot, Neutralization | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| beta chemokine exodus-1, Beta-chemokine exodus-1, CC chemokine LARC, C-C motif chemokine 20, chemokine (C-C motif) ligand 20, CKb4, exodus-1, LARC, LARCLiver and activation-regulated chemokine, MIP3 alpha, MIP-3 alpha, MIP-3a, MIP-3-alpha, MIP3AMacrophage inflammatory protein 3 alpha, SCYA20Small-inducible cytokine A20, small inducible cytokine subfamily A (Cys-Cys), member 20, ST38 | |
| CCL20 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title