Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CCL21/6Ckine Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC457R100UG
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC457R100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC457R100UG Supplier R&D Systems Supplier No. IC457R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CCL21/6Ckine Monoclonal specifically detects CCL21/6Ckine in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen CCL21/6Ckine
Applications Flow Cytometry
Classification Monoclonal
Clone 59106
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 647
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P84444
Gene Alias 6Ckine, 6CkineSmall-inducible cytokine A21, Beta-chemokine exodus-2, chemokine (C-C motif) ligand 21, CKb9, exodus-2, member 21, SCYA21, SCYA21MGC34555, secondary lymphoid tissue chemokine, SLC, SLCSecondary lymphoid-tissue chemokine, TCA-4
Gene Symbols Ccl21a
Host Species Rat
Immunogen E. coli-derived recombinant mouse CCL21/6Ckine Ser24-Gly133 Accession # P84444
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6366
Test Specificity Detects mouse CCL21/6Ckine in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) 6Ckine, rhMIP-1 alpha , rhMIP-1 beta , recombinant mouse (rm) MIP-1 alpha , rmMIP-1 beta , rhMIP-1 delta , rmMIP-1 gamma , rmMIP-2, rhMIP-3 alpha , rmMIP-3 alpha , rhMIP-3 beta , rmMIP-3 beta , rhTeck, or rmTeck.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.