Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CCL22/MDC Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF439SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF439SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF439SP Supplier R&D Systems Supplier No. AF439SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 8 publications

CCL22/MDC Polyclonal specifically detects CCL22/MDC in Mouse samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen CCL22/MDC
Applications Western Blot, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Neutralization 0.3-1.0 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9QZU2
Gene Alias A-152E5.1, ABCD-1, CC chemokine STCP-1, C-C motif chemokine 22, chemokine (C-C motif) ligand 22, DC/B-CK, Macrophage-derived chemokine, MDC, MDCStimulated T-cell chemotactic protein 1, MGC34554, SCYA22MDC(1-69), small inducible cytokine A22, small inducible cytokine subfamily A (Cys-Cys), member 22, Small-inducible cytokine A22, STCP-1, stimulated T cell chemotactic protein 1
Gene Symbols CCL22
Host Species Goat
Immunogen E. coli-derived recombinant mouse CCL22/MDC Gly25-Ser92 Accession # Q9QZU2
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6367
Test Specificity Detects mouse CCL22/MDC in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human CCL22/MDC is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.