Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CCL22/MDC Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4391100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4391100 100 μg
MAB4391SP 25 μg
MAB4391500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB4391100 Supplier R&D Systems Supplier No. MAB4391100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 4 publications

CCL22/MDC Monoclonal specifically detects CCL22/MDC in Mouse samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen CCL22/MDC
Applications Neutralization, ELISA
Classification Monoclonal
Clone 158132
Conjugate Unconjugated
Dilution Neutralization 0.4-2.0 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O88430
Gene Alias A-152E5.1, ABCD-1, CC chemokine STCP-1, C-C motif chemokine 22, chemokine (C-C motif) ligand 22, DC/B-CK, Macrophage-derived chemokine, MDC, MDCStimulated T-cell chemotactic protein 1, MGC34554, SCYA22MDC(1-69), small inducible cytokine A22, small inducible cytokine subfamily A (Cys-Cys), member 22, Small-inducible cytokine A22, STCP-1, stimulated T cell chemotactic protein 1
Gene Symbols CCL22
Host Species Rat
Immunogen E. coli-derived recombinant mouse CCL22/MDC Gly25-Ser92 Accession # O88430
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6367
Test Specificity Detects mouse CCL22/MDC in ELISAs and Western blots. In Western blots, this antibody shows 25% cross-reactivity with recombinant viral MIP-II and no cross-reactivity with rmCCL1, 2, 3, 4, 6, 7, CCL9/10/MIP-1 gamma , 11, 12, 19, 20, 21, 22, 24, 25, 27, rhCCL1, 2, 3, 4, 5, 7, 8, 11, 13, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, 28, or rrCCL20.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.