Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CCL22/MDC Antibody, R&D Systems™



Goat Polyclonal Antibody has been used in 8 publications
Supplier: R&D Systems AF439
This item is not returnable.
View return policy
Description
CCL22/MDC Polyclonal specifically detects CCL22/MDC in Mouse samples. It is validated for Western Blot, Neutralization.Specifications
CCL22/MDC | |
Polyclonal | |
Unconjugated | |
Q9QZU2 | |
CCL22 | |
E. coli-derived recombinant mouse CCL22/MDC Gly25-Ser92 Accession # Q9QZU2 | |
100 μg | |
Primary | |
Detects mouse CCL22/MDC in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human CCL22/MDC is observed. | |
Mouse | |
IgG |
Western Blot, Neutralization | |
LYOPH | |
Western Blot 0.1 ug/mL, Neutralization 0.3-1.0 ug/mL | |
A-152E5.1, ABCD-1, CC chemokine STCP-1, C-C motif chemokine 22, chemokine (C-C motif) ligand 22, DC/B-CK, Macrophage-derived chemokine, MDC, MDCStimulated T-cell chemotactic protein 1, MGC34554, SCYA22MDC(1-69), small inducible cytokine A22, small inducible cytokine subfamily A (Cys-Cys), member 22, Small-inducible cytokine A22, STCP-1, stimulated T cell chemotactic protein 1 | |
Goat | |
Affinity Purified | |
RUO | |
6367 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction