Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CCL25/TECK Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
$200.50 - $2020.00
Specifications
Antigen | CCL25/TECK |
---|---|
Clone | 89818 |
Dilution | Western Blot 1 ug/mL, Neutralization 1.5-6.0 ug/mL |
Applications | Western Blot, Neutralization |
Classification | Monoclonal |
Description
CCL25/TECK Monoclonal specifically detects CCL25/TECK in Mouse samples. It is validated for Western Blot, Neutralization.Specifications
CCL25/TECK | |
Western Blot 1 ug/mL, Neutralization 1.5-6.0 ug/mL | |
Monoclonal | |
Rat | |
Mouse | |
O35903.1 | |
6370 | |
E. coli-derived recombinant mouse CCL25/TECK Gln24-Asn144 Accession # O35903.1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse CCL25/TECK in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) CCL1, 2, 3, 4, 5, 6, 7, 9, 11, 12, 17, 19, 20, 21, 22 or 24, recombinant human (rh) CCL2, 3, 5, 7, 8, 11, 13, 14, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25 or 26, or recombinant rat (rr) CCL20 is observed. |
89818 | |
Western Blot, Neutralization | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C-C motif chemokine 25, chemokine (C-C motif) ligand 25, Chemokine TECK, Ck beta-15, Ckb15, SCYA25TECKvar, small inducible cytokine subfamily A (Cys-Cys), member 25, Small-inducible cytokine A25, TECK, TECKMGC150327, thymus expressed chemokine, Thymus-expressed chemokine | |
CCL25 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title