Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CCL25/TECK Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4811
Change view
Click to view available options
Quantity:
25 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4811 500 μg
MAB4811SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB4811 Supplier R&D Systems Supplier No. MAB4811
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 3 publications

CCL25/TECK Monoclonal specifically detects CCL25/TECK in Mouse samples. It is validated for Western Blot, Immunohistochemistry, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen CCL25/TECK
Applications Western Blot, Immunohistochemistry, ELISA
Classification Monoclonal
Clone 89827
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O35903.1
Gene Alias C-C motif chemokine 25, chemokine (C-C motif) ligand 25, Chemokine TECK, Ck beta-15, Ckb15, SCYA25TECKvar, small inducible cytokine subfamily A (Cys-Cys), member 25, Small-inducible cytokine A25, TECK, TECKMGC150327, thymus expressed chemokine, Thymus-expressed chemokine
Gene Symbols CCL25
Host Species Rat
Immunogen E. coli-derived recombinant mouse CCL25/TECK Gln24-Asn144 Accession # O35903.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6370
Test Specificity Detects mouse CCL25/TECK in ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human CCL1, 2, 3, 4, 5, 7, 8, 11, 13, 14, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, recombinant mouse CCL1, 2, 3, 4, 6, 7, 9, 11, 12, 19, 20, 21, 22, 24, and recombinant rat CCL20 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.