Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CCL27/CTACK Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
Supplier: R&D Systems MAB725
This item is not returnable.
View return policy
Description
CCL27/CTACK Monoclonal specifically detects CCL27/CTACK in Mouse samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
CCL27/CTACK | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ALP, C-C motif chemokine 27, chemokine (C-C motif) ligand 27, CTACK, CTACKSmall-inducible cytokine A27, CTAK, cutaneous T-cell attracting chemokine, Cutaneous T-cell-attracting chemokine, ESKINE, IL-11 Ralpha-locus chemokine, ILC, ILCCC chemokine ILC, PESKY, SCYA27IL-11 R-alpha-locus chemokine, Skinkine, small inducible cytokine subfamily A (Cys-Cys), member 27 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10850 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Neutralization, ELISA | |
68623 | |
Neutralization 2.5-10 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
NP_035466 | |
Ccl27a | |
E. coli-derived recombinant human CCL27/CTACK Leu26-Asn120 Accession # NP_035466 | |
500 μg | |
Primary | |
Detects mouse CCL27/CTACK in direct ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant cotton rat CCL5, recombinant human CCL1, 2, 3, 4, 5, 7, 8, 11, 13, 14, 15, 16, 17, 18, 19, 20, 21, 22, 23, 25, 27, 28, recombinant mouse CCL1, 2, 3, 4, 6, 7, 9/10/MIP-1 gamma , 11, 12, 19, 21, 22, 25, or recombinant rat CCL20. | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction