Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CCL27/CTACK Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF725
This item is not returnable.
View return policy
Description
CCL27/CTACK Polyclonal specifically detects CCL27/CTACK in Mouse samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
CCL27/CTACK | |
Polyclonal | |
Biotin | |
NP_035466 | |
Ccl27a | |
E. coli-derived recombinant human CCL27/CTACK Leu26-Asn120 Accession # NP_035466 | |
50 μg | |
Primary | |
Detects mouse CCL27/CTACK in ELISAs and Western blots. In sandwich immunoassays, less than 0.2% cross-reactivity with recombinant human CTACK, recombinant mouse (rm) MIP-3 beta , rmTECK, rmGDF-6, and rmFractalkine is observed. | |
Mouse | |
IgG |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
ALP, C-C motif chemokine 27, chemokine (C-C motif) ligand 27, CTACK, CTACKSmall-inducible cytokine A27, CTAK, cutaneous T-cell attracting chemokine, Cutaneous T-cell-attracting chemokine, ESKINE, IL-11 Ralpha-locus chemokine, ILC, ILCCC chemokine ILC, PESKY, SCYA27IL-11 R-alpha-locus chemokine, Skinkine, small inducible cytokine subfamily A (Cys-Cys), member 27 | |
Goat | |
Affinity Purified | |
RUO | |
10850 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction