Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CCL3/MIP-1 alpha Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4501
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4501 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB4501 Supplier R&D Systems Supplier No. MAB4501
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CCL3/MIP-1 alpha Monoclonal specifically detects CCL3/MIP-1 alpha in Mouse samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen CCL3/MIP-1 alpha
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 756613
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P10855
Gene Alias C-C motif chemokine 3, MIP1-(a), AI323804, chemokine (C-C motif) ligand 3, G0S19-1, LD78a, LD78alpha, MIP1 alpha, MIP-1 alpha, Mip1a, MIP-1alpha, MIP1-alpha, PAT 464.1, SCYA3, SIS-beta
Gene Symbols CCL3
Host Species Rat
Immunogen E. coli-derived recombinant mouse CCL3/MIP-1 alpha Ala24-Ala92 Accession # P10855
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6348
Test Specificity Detects mouse CCL3/MIP-1 alpha in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant rat CCL3 and 50% cross-reactivity with recombinant human CCL3 is observed. No cross-reactivity with recombinant canine CCL3, recombinant cotton rat CCL3, recombinant mouse (rm) CCL4/MIP-1 beta, or rmCCL5 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.