Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CCL4/MIP-1 beta Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB451500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB451500 500 μg
MAB451SP 25 μg
MAB451100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB451500 Supplier R&D Systems Supplier No. MAB451500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 5 publications

CCL4/MIP-1 beta Monoclonal specifically detects CCL4/MIP-1 beta in Mouse samples. It is validated for Neutralization.

Specifications

Antigen CCL4/MIP-1 beta
Applications Neutralization
Classification Monoclonal
Clone 46907
Conjugate Unconjugated
Dilution Neutralization 1-3 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q5QNV9
Gene Alias ACT2, ACT-2, AT744.1, C-C motif chemokine 4, chemokine (C-C motif) ligand 4, Exodus-3, G-26, G-26 T-lymphocyte-secreted protein, HC21, LAG1, LAG-1, Lymphocyte activation gene 1 protein, Macrophage inflammatory protein 1-beta, MIP1 beta, MIP-1 beta, MIP1B, MIP1B1, MIP-1-beta, MIP-1-beta(1-69), PAT 744, Protein H400, SCYA2, SCYA4, secreted protein G-26, SIS-gamma, small inducible cytokine A4 (homologous to mouse Mip-1b), Small-inducible cytokine A4, T-cell activation protein 2
Gene Symbols CCL4
Host Species Rat
Immunogen E. coli-derived recombinant mouse CCL4/MIP-1 beta Ala24-Asn92 Accession # Q5QNV9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6351
Test Specificity Detects mouse CCL4/MIP-1 beta in ELISAs. Does not cross-react with recombinant human (rh) CCL1, 2, 4/MIP-1 alpha , 4/MIP-1 beta , 5, 7, 8, 11, 13, 14, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, 26, rmCCL1, 2, 4/MIP-1 alpha , 4/MIP-1 beta , 5, 6, 7, 9/10, 11, 12, 17, 19, 20, 21, 22, 24, 25, or rrCCL20.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.