Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CCL6/C10 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems IC487V100UG
This item is not returnable.
View return policy
Description
CCL6/C10 Monoclonal specifically detects CCL6/C10 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
CCL6/C10 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P27784 | |
Rat | |
100 μg | |
Primary | |
Detects mouse CCL6/C10 in Western blots. In Western blots, no cross-reactivity with recombinant human CCL1, 2, 3, 4, 5, 7, 8, 11, 13, 14, 15, 16, 17, 18, 19, 21, 22, 23, 24, 25, 28, recombinant mouse CCL3, 4, 9, 11, 12, 19, 20, 21, 22, 24, 25, 28, or recombinant rat CCL20 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Flow Cytometry | |
262016 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
Ccl6 | |
E. coli-derived recombinant mouse CCL6/C10 Gly22-Ala116 Accession # P27784 | |
RUO | |
20305 | |
Mouse | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction