Learn More
Mouse CCL6/C10 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | CCL6/C10 |
| Applications | Western Blot, Neutralization |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL, Neutralization 70-100 ug/mL |
| Gene Accession No. | P27784 |
| Gene Symbols | Ccl6 |
| Host Species | Goat |
| Immunogen | E. coli-derived recombinant mouse CCL6/C10 Gly22-Ala116 Accession # P27784 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.