Learn More
Mouse CCL6/C10 Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody has been used in 3 publications
Supplier:  R&D Systems AF487
Catalog No. AF487
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
CCL6/C10 Polyclonal specifically detects CCL6/C10 in Mouse samples. It is validated for Western Blot, Neutralization.
Specifications
Specifications
CCL6/C10
Polyclonal
Unconjugated
P27784
Goat
Affinity Purified
RUO
20305
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot, Neutralization
LYOPH
Western Blot 0.1 ug/mL, Neutralization 2-10 ug/mL
Ccl6
E. coli-derived recombinant mouse CCL6/C10 Gly22-Ala116 Accession # P27784
100 μg
Primary
Detects CCL6/C10 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 20% cross-reactivity with recombinant mouse MIP-1 gamma is observed and less than 5% cross-reactivity with recombinant human MIP-1 delta is observed.
Mouse
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.