Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CCL6/C10 Biotinylated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems BAM4872
This item is not returnable.
View return policy
Description
CCL6/C10 Monoclonal specifically detects CCL6/C10 in Mouse samples. It is validated for ELISA Detection (Matched Antibody Pair).Specifications
CCL6/C10 | |
Monoclonal | |
LYOPH | |
ELISA Detection (Matched Antibody Pair) 0.5-2 ug/mL | |
P27784 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
20305 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
ELISA | |
262008 | |
Biotin | |
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative | |
Ccl6 | |
E. coli-derived recombinant mouse CCL6/C10 Gly22-Ala116 Accession # P27784 | |
250 μg | |
Primary | |
Detects mouse CCL6/C10 in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant mouse (rm) CCL3, rmCCL4, or rmCCL9/10 is observed. | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction