Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CCL8/MCP-2 Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. MAB790500 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB790500 500 μg
MAB790SP 25 μg
MAB790100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB790500 Supplier R&D Systems Supplier No. MAB790500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 2 publications

CCL8/MCP-2 Monoclonal specifically detects CCL8/MCP-2 in Mouse samples. It is validated for Immunohistochemistry∣Immunohistochemistry (Paraffin)∣Neutralization∣Western Blot.

Specifications

Antigen CCL8/MCP-2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Neutralization, Western Blot
Classification Monoclonal
Clone 146123
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9Z121
Gene Alias C-C motif chemokine 8, chemokine (C-C motif) ligand 8, HC14, MCP2, MCP-2, member 8 (monocyte chemotactic protein 2), monocyte chemoattractant protein 2, SCYA10, small-inducible cytokine A8
Gene Symbols CCL8
Host Species Rat
Immunogen E. coli-derived recombinant mouse CCL8/MCP-2 Glu20-Pro97 Accession # Q9Z121
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6355
Test Specificity Detects mouse CCL8/MCP-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant cotton rat CCL3, 4, 5, recombinant human CCL1, 2, 3, 4, 5, 7, 8, 11, 13, 14, 15, 16, 17, 18, 19, 20, 21, 22, 23, 24, 25, 26, 27, 28, recombinant mouse CCL1, 3, 4, 5, 6, 7, 9/10MIP-1 gamma , 11, 12, 17, 19, 20, 21, 22, 24, 25, 27, or recombinant rat CCL20 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.