Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CCL8/MCP-2 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF790
This item is not returnable.
View return policy
Description
CCL8/MCP-2 Polyclonal specifically detects CCL8/MCP-2 in Mouse samples. It is validated for Western Blot.Specifications
CCL8/MCP-2 | |
Polyclonal | |
Biotin | |
Q9Z121 | |
CCL8 | |
E. coli-derived recombinant mouse CCL8/MCP-2 Glu20-Pro97 Accession # Q9Z121 | |
50 μg | |
Primary | |
Detects mouse CCL8/MCP-2 in Western blots. In Western blots, less than 1% cross-reactivity with Brucella melitensis BcsP31 is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
C-C motif chemokine 8, chemokine (C-C motif) ligand 8, HC14, MCP2, MCP-2, member 8 (monocyte chemotactic protein 2), monocyte chemoattractant protein 2, SCYA10, small-inducible cytokine A8 | |
Goat | |
Affinity Purified | |
RUO | |
6355 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction