Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
The Mouse CCL9/10/MIP-1 gamma DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse CCL9/10/MIP-1 gamma.
Specifications
Specifications
| Quantity | 15 x 96-well Plates |
| Conjugate | Biotin |
| Assay Range | 7.81 to 500 pg/mL |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 20308 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
