Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CCR1 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems MAB5986
This item is not returnable.
View return policy
Description
CCR1 Monoclonal specifically detects CCR1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
CCR1 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
P51675 | |
CCR1 | |
E. coli-derived recombinant mouse CCR1 Extracellular regions (aa 1-34, aa 92-107, aa 172-192, aa 265-281) Accession # P51675 | |
100 μg | |
Primary | |
Detects mouse CCR1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse CCR4, 5, or 8 is observed. | |
Mouse | |
Purified |
Western Blot, Flow Cytometry, CyTOF | |
643854 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C-C chemokine receptor type 1, CC-CKR-1, CCR1, CCR-1, CD191, CD191 antigen, chemokine (C-C motif) receptor 1, CKR1, CKR-1, CMKBR1SCYAR1, CMKR1, HM145C-C CKR-1, LD78 receptor, Macrophage inflammatory protein 1-alpha receptor, MIP-1alpha-R, MIP1aR, RANTES receptor, RANTES-R | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1230 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction