Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CCR1 PE-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
$210.00 - $479.50
Specifications
| Antigen | CCR1 |
|---|---|
| Clone | 643854 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB5986P025
![]() |
R&D Systems
FAB5986P025 |
25 Tests |
Each for $210.00
|
|
|||||
FAB5986P100
![]() |
R&D Systems
FAB5986P100 |
100 Tests |
Each for $479.50
|
|
|||||
Description
CCR1 Monoclonal specifically detects CCR1 in Mouse samples. It is validated for Flow Cytometry.Specifications
| CCR1 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| Mouse | |
| C-C chemokine receptor type 1, CC-CKR-1, CCR1, CCR-1, CD191, CD191 antigen, chemokine (C-C motif) receptor 1, CKR1, CKR-1, CMKBR1SCYAR1, CMKR1, HM145C-C CKR-1, LD78 receptor, Macrophage inflammatory protein 1-alpha receptor, MIP-1alpha-R, MIP1aR, RANTES receptor, RANTES-R | |
| CCR1 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects mouse CCR1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse CCR4, 5, or 8 is observed. |
| 643854 | |
| Flow Cytometry | |
| R-PE | |
| Rat | |
| P51675 | |
| 1230 | |
| E. coli-derived recombinant mouse CCR1 Extracellular regions (aa 1-34, aa 92-107, aa 172-192, aa 265-281) Accession # P51675 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title