Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CCR10 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2815R100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2815R100 100 μg
MAB2815RSP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB2815R100 Supplier R&D Systems Supplier No. MAB2815R100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CCR10/GPR2 Monoclonal specifically detects CCR10/GPR2 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen CCR10/GPR2
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 248918R
Conjugate Unconjugated
Dilution Flow Cytometry 0.25μg/106 cells, CyTOF-ready
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
Gene Accession No. NP_031747
Gene Alias CC chemokine receptor 10, C-C chemokine receptor type 10, C-C CKR-10, CC-CKR-10, CCR10, CCR-10, chemokine (C-C motif) receptor 10, G protein-coupled receptor 2, GPR2, GPR2G-protein coupled receptor 2
Host Species Rat
Immunogen C6 rat glioma cell line transfected with mouse CCR10 Met1-Asn362 Accession # NP_031747
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 2826
Test Specificity Detects mouse CCR10 in direct ELISAs.
Reconstitution Reconstitute at 0.5mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.