Learn More
Mouse CCR10 PerCP-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | CCR10/GPR2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 248918 |
| Conjugate | PerCP |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Gene Accession No. | NP_031747 |
| Gene Alias | CC chemokine receptor 10, C-C chemokine receptor type 10, C-C CKR-10, CC-CKR-10, CCR10, CCR-10, chemokine (C-C motif) receptor 10, G protein-coupled receptor 2, GPR2, GPR2G-protein coupled receptor 2 |
| Gene Symbols | CCR10 |
| Host Species | Rat |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.