Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CCR2 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $526.00
Specifications
Antigen | CCR2 |
---|---|
Clone | 475303 |
Dilution | Immunocytochemistry 8-25 μg/mL |
Applications | Immunocytochemistry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB55382SP
![]() |
R&D Systems
MAB55382SP |
25 μg |
Each for $200.50
|
|
|||||
MAB55382100
![]() |
R&D Systems
MAB55382100 |
100 μg |
Each for $526.00
|
|
|||||
Description
CCR2 Monoclonal specifically detects CCR2 in Mouse samples. It is validated for Immunocytochemistry.Specifications
CCR2 | |
Immunocytochemistry 8-25 μg/mL | |
Monoclonal | |
Rat | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
729230 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
475303 | |
Immunocytochemistry | |
Unconjugated | |
Mouse | |
C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2, CC-CKR-2CKR2B, CCR2, CCR-2, CCR2A, CCR2B, CD192, CD192 antigen, chemokine (C-C motif) receptor 2, CKR2, CKR2A, CMKBR2, CMKBR2MGC111760, FLJ78302, MCP-1 receptor, MCP-1-R, MCP-1-RMGC103828, MGC168006, Monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor | |
L1.2 mouse pro-B cell line transfected with mouse CCR2 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse CCR2 in immunocytochemistry. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title