Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CCR2 Fluorescein-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB5538F025
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB5538F025 25 Tests
FAB5538F100 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. FAB5538F025 Supplier R&D Systems Supplier No. FAB5538F025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 12 publications

CCR2 Monoclonal specifically detects CCR2 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen CCR2
Applications Flow Cytometry
Classification Monoclonal
Clone 475301
Conjugate Fluorescein
Dilution Flow Cytometry 10 uL/10^6 cells
Gene Alias C-C chemokine receptor type 2, C-C CKR-2, CC-CKR-2, CC-CKR-2CKR2B, CCR2, CCR-2, CCR2A, CCR2B, CD192, CD192 antigen, chemokine (C-C motif) receptor 2, CKR2, CKR2A, CMKBR2, CMKBR2MGC111760, FLJ78302, MCP-1 receptor, MCP-1-R, MCP-1-RMGC103828, MGC168006, Monocyte chemoattractant protein 1 receptor, monocyte chemotactic protein 1 receptor
Gene Symbols CCR2
Host Species Rat
Immunogen L1.2 mouse pro-B cell line transfected with mouse CCR2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Primary or Secondary Primary
Gene ID (Entrez) 729230
Test Specificity Detects mouse CCR2 transfectants but not the parental cell line in flow cytometry. Does not detect HEK293 human embryonic kidney cell line transfected with mouse CCR5 in flow cytometry.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.