Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CCR3 PE-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 11 publications
$162.90 - $456.00
Specifications
Antigen | CCR3 |
---|---|
Clone | 83101 |
Dilution | Flow Cytometry 0.25-1 μg/106 cells |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FAB729P025U
![]() |
R&D Systems
FAB729P025UG |
25 μg |
Each for $162.90
|
|
|||||
FAB729P100U
![]() |
R&D Systems
FAB729P100UG |
100 μg |
Each for $456.00
|
|
|||||
Description
CCR3 Monoclonal specifically detects CCR3 in Mouse samples. It is validated for Flow Cytometry.Specifications
CCR3 | |
Flow Cytometry 0.25-1 μg/106 cells | |
Monoclonal | |
Supernatant | |
Mouse | |
P51678 | |
1232.0 | |
Y3 rat myeloid cell line transfected with mouse CCR3, Met1-Phe359, Accession No. P51678 | |
Primary | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. |
83101 | |
Flow Cytometry | |
R-PE | |
Rat | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
b-chemokine receptor, CC chemokine receptor 3, C-C chemokine receptor type 3, C-C CKR-3, CCR3, CCR-3, CD193, CD193 antigen, chemokine (C-C motif) receptor 3, CKR3CC-CKR-3CMKBR3CD193, eosinophil CC chemokine receptor 3, Eosinophil eotaxin receptor, MGC102841 | |
CCR3 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Detects mouse CCR3. Specifically stains mouse CCR3 transfectants but not the parental cell lines. Does not cross-react with Y3 cells transfected with human CCR3. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title