Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CCR9 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2160100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2160100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB2160100 Supplier R&D Systems Supplier No. MAB2160100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 8 publications

CCR9 Monoclonal specifically detects CCR9 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen CCR9
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 242503
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9WUT7
Gene Alias C-C chemokine receptor type 9, C-C CKR-9, CC-CKR-9, CCR9, CCR-9, CD199, CDw199, CDw199 antigen, chemokine (C-C motif) receptor 9, G protein-coupled receptor 28, GPR28, GPR-9-6G-protein coupled receptor 28
Gene Symbols CCR9
Host Species Rat
Immunogen Y3 rat myeloid cell line transfected with mouse CCR9 Met1-Leu369 Accession # Q9WUT7
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10803
Test Specificity Detects mouse CCR9 in flow cytometry. Stains mouse CCR9-transfected cells but not irrelevant transfectants.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.