Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CCRL2/LCCR Alexa Fluor 405-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB5519V100UG
This item is not returnable.
View return policy
Description
CCRL2/LCCR Monoclonal specifically detects CCRL2/LCCR in Mouse samples. It is validated for Flow Cytometry.Specifications
CCRL2/LCCR | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
O35457 | |
Rat | |
100 μg | |
Primary | |
Detects mouse CCRL2/LCCR in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Flow Cytometry | |
498321 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
Ccrl2 | |
RBL-1 rat basophilic leukemia cell line transfected with mouse CCRL2/LCCR Accession # O35457 | |
RUO | |
9034 | |
Mouse | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction