Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CD11b/Integrin alpha M Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$200.50 - $467.50
Specifications
Antigen | CD11b |
---|---|
Clone | 1250D |
Dilution | Western Blot 0.5 ug/mL, Simple Western 5 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB11242SP
![]() |
R&D Systems
MAB11242SP |
25 μg |
Each for $200.50
|
|
|||||
MAB11242100
![]() |
R&D Systems
MAB11242100 |
100 μg |
Each for $467.50
|
|
|||||
Description
CD11b Monoclonal antibody specifically detects CD11b in Mouse samples. It is validated for Western Blot, Western BlotSpecifications
CD11b | |
Western Blot 0.5 ug/mL, Simple Western 5 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, Cell surface glycoprotein MAC-1 subunit alpha, Complement component 3 receptor 3 subunit, CR-3 alpha chain, CR3A, CR3AMGC117044, Integrin alpha M, integrin alpha-M, integrin, alpha M (complement component 3 receptor 3 subunit), ITGAM, Leukocyte adhesion receptor MO1, MAC-1, MAC1A, macrophage antigen alpha polypeptide, MO1A, Neutrophil adherence receptor, neutrophil adherence receptor alpha-M subunit, SLEB6 | |
MouseCD11b/Integrin alpha M synthetic peptide, Accession # P11215 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1250D | |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse | |
P11215 | |
3684 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title