Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CD11b/Integrin alpha M Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 17 publications
$212.05 - $2160.65
Specifications
| Antigen | CD11b |
|---|---|
| Clone | M1/70 |
| Concentration | LYOPH |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry 8-25 ug/mL, CyTOF-reported |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, CyTOF |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1124SP
![]() |
R&D Systems
MAB1124SP |
25 μg |
Each for $212.05
|
|
|||||
MAB1124100
![]() |
R&D Systems
MAB1124100 |
100 μg |
Each for $579.81
|
|
|||||
MAB1124500
![]() |
R&D Systems
MAB1124500 |
500 μg |
Each for $2,160.65
|
|
|||||
Description
CD11b Monoclonal specifically detects CD11b in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, CyTOF-reported.Specifications
| CD11b | |
| LYOPH | |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, CyTOF | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 3684 | |
| Con A-activated C57BL/10 splenocytes | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse CD11b/Integrin alpha M. Cross-reaction with human CD11b/Integrin alpha M has been reported (1, 2). |
| M1/70 | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry 8-25 ug/mL, CyTOF-reported | |
| Monoclonal | |
| Rat | |
| Mouse | |
| antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, Cell surface glycoprotein MAC-1 subunit alpha, Complement component 3 receptor 3 subunit, CR-3 alpha chain, CR3A, CR3AMGC117044, Integrin alpha M, integrin alpha-M, integrin, alpha M (complement component 3 receptor 3 subunit), ITGAM, Leukocyte adhesion receptor MO1, MAC-1, MAC1A, macrophage antigen alpha polypeptide, MO1A, Neutrophil adherence receptor, neutrophil adherence receptor alpha-M subunit, SLEB6 | |
| ITGAM | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title