Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CD14 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications
Supplier: R&D Systems MAB982
This item is not returnable.
View return policy
Description
CD14 Monoclonal specifically detects CD14 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
CD14 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD14, CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
929 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot, Flow Cytometry, CyTOF | |
159010 | |
Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
P10810 | |
CD14 | |
Mouse myeloma cell line NS0-derived recombinant mouse CD14 Ala18-Pro345 (predicted) Accession # P10810 | |
500 μg | |
Primary | |
Detects mouse CD14 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant human CD14 is observed. | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction