Learn More
Mouse CD151 PE-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | CD151 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 455807 |
| Conjugate | R-PE |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Formulation | Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide |
| Gene Accession No. | BAA22447 |
| Gene Alias | CD151, CD151 antigen (Raph blood group), CD151 antigenplatelet surface glycoprotein gp27, CD151 molecule (Raph blood group), GP27, Membrane glycoprotein SFA-1, MER2, PETA3, PETA-3, PETA-3SFA-1, platelet-endothelial cell tetraspan antigen 3, Platelet-endothelial tetraspan antigen 3, RAPH, SFA1, SFA-1, tetraspanin-24, tspan-24, TSPAN24, TSPAN24hemidesmosomal tetraspanin CD151 |
| Gene Symbols | CD151 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.