Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CD161/NK1.1 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB7614V100UG
This item is not returnable.
View return policy
Description
CD161/NK1.1 Monoclonal specifically detects CD161/NK1.1 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD161/NK1.1 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P27814 | |
KLRB1C | |
Mouse myeloma cell line NS0-derived recombinant mouse CD161/NK1.1 Val62-Ser220 Accession # P27814 | |
RUO | |
17059 | |
Mouse | |
IgG2a |
Flow Cytometry | |
694370 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD161, CD161 antigen, CD161c, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, Klrb1c, Ly55c, Ly-59, Natural killer cell surface protein P1A, NKR-P1, NKR-P1AMGC138614, NKRP1ANKR | |
Rat | |
100 μg | |
Primary | |
Detects mouse CD161/NK1.1 expressed specifically in the C57/B6 mouse strain. No detection of CD161/NK1.1 was observed in the BALB/c mouse strain. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse (rm) KLRB-1B is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction