Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CD161/NK1.1 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB76141A100UG
This item is not returnable.
View return policy
Description
CD161/NK1.1 Monoclonal specifically detects CD161/NK1.1 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD161/NK1.1 | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. | |
CD161, CD161 antigen, CD161c, CLEC5BC-type lectin domain family 5 member B, HNKR-P1a, killer cell lectin-like receptor subfamily B member 1, killer cell lectin-like receptor subfamily B, member 1, Klrb1c, Ly55c, Ly-59, Natural killer cell surface protein P1A, NKR-P1, NKR-P1AMGC138614, NKRP1ANKR | |
NK-1+ cells from mouse spleen and bone marrow, Accession # AAH61168 | |
100 μg | |
Primary | |
Mouse | |
Purified |
Flow Cytometry | |
PK136 | |
Flow Cytometry 0.25-1 ug/106 cells | |
AAH61168 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
17059 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction