Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CD177 Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB8186G025
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB8186G025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB8186G025 Supplier R&D Systems Supplier No. FAB8186G025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

CD177 Monoclonal specifically detects CD177 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen CD177
Applications Flow Cytometry
Classification Monoclonal
Clone 1171A
Conjugate Alexa Fluor 488
Dilution Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. Q8R2S8
Gene Alias CD177, CD177 antigenNB1 GP, CD177 molecule, HNA-2a, HNA2Acell surface receptor, Human neutrophil alloantigen 2a, NB1 GP, NB1PRV-1, polycythemia rubra vera 1, Polycythemia rubra vera protein 1, PRV1, PRV1NB1 glycoprotein
Gene Symbols CD177
Host Species Rabbit
Immunogen Human embryonic kidney cell line HEK293-derived recombinant mouse CD177 Accession # Q8R2S8
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57126
Test Specificity Detects mouse CD177 in direct ELISAs.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.