Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CD1d1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF4884
This item is not returnable.
View return policy
Description
CD1d1 Polyclonal specifically detects CD1d1 in Mouse samples. It is validated for Western Blot.Specifications
CD1d1 | |
Polyclonal | |
Unconjugated | |
NP_031665 | |
Cd1d1 | |
Mouse myeloma cell line NS0-derived recombinant mouse CD1d1 Gln22-Gly305 Accession # NP_031665 | |
100 μg | |
Primary | |
Detects mouse CD1d1 in direct ELISAs and Western blots. In direct ELISAs, approximately 70% cross-reactivity with recombinant mouse CD1d2 is observed, and approximately 15% cross-reactivity with recombinant human CD1d1 is observed. | |
Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.5 ug/mL | |
AI747460, CD1.1, CD1A, Cd1d, Ly-38 | |
Sheep | |
Affinity Purified | |
RUO | |
912 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction