Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CD200R1 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF2554
This item is not returnable.
View return policy
Description
CD200R1 Polyclonal specifically detects CD200R1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry.Specifications
CD200R1 | |
Polyclonal | |
Biotin | |
BAE42266 | |
CD200R1 | |
Mouse myeloma cell line NS0-derived recombinant mouse CD200 R1 Thr26-Pro238 Accession # BAE42266 | |
50 μg | |
Primary | |
Detects mouse CD200 R1 in Western blots. In Western blots, less than 5% cross-reactivity with recombinant human CD200 R is observed. | |
Mouse | |
IgG |
Western Blot, Flow Cytometry, Immunocytochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL | |
CD200 R1, CD200 receptor 1, CD200RMOX2Rcell surface glycoprotein CD200 receptor 1, Cell surface glycoprotein OX2 receptor 1, cell surface glycoprotein receptor CD200, CRTR2, HCRTR2, MOX2 receptor, MOX2R, OX2RCD200 cell surface glycoprotein receptor | |
Goat | |
Affinity Purified | |
RUO | |
131450 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Store the unopened product at -20°C to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction