Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CD23/Fc epsilon RII Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB6900SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB6900SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB6900SP Supplier R&D Systems Supplier No. MAB6900SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CD23/Fc epsilon RII Monoclonal specifically detects CD23/Fc epsilon RII in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen CD23/Fc epsilon RII
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 691632
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P20693
Gene Alias BLAST-2, CD23, CD23A, CD23CD23 antigen, CLEC4J, CLEC4JC-type lectin domain family 4 member J, C-type lectin domain family 4, member J, Fc epsilon RII, Fc fragment of IgE, low affinity II, receptor for (CD23), FCE2Fc fragment of IgE, low affinity II, receptor for (CD23A), fc-epsilon-RII, FCER2, Fcer2a, FceRII, IGEBF, Immunoglobulin E-binding factor, low affinity immunoglobulin epsilon Fc receptor, Ly-42, Lymphocyte IgE receptor
Gene Symbols Fcer2a
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CD23/Fc epsilon RII Glu50-Pro331 Accession # P20693
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2208
Test Specificity Detects mouse CD23/Fc epsilon RII in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) Fc epsilon RIA or rhFc epsilon RIG is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.