Learn More
Mouse CD25/IL-2R alpha Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody
Supplier:  R&D Systems FAB9164T025
Catalog No. FAB9164T025
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
CD25/IL-2R alpha Monoclonal specifically detects CD25/IL-2R alpha in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD25/IL-2R alpha
Monoclonal
Alexa Fluor 594
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
IL2RA
B6.1 mouse cytotoxic T cell line
25 Tests
Primary
Detects mouse CD25/IL-2 R alpha in flow cytometry.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG1
Flow Cytometry
PC61.5.3
Flow Cytometry 5 uL/10^6 cells
CD25, CD25 antigen, IDDM10, IL-2 R alpha, IL-2 receptor subunit alpha, IL2R, IL2R alpha, IL-2R subunit alpha, IL-2Ra, IL2RA, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
3559
Mouse
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.