Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CD25/IL-2R alpha APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB9164A025
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB9164A025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB9164A025 Supplier R&D Systems Supplier No. FAB9164A025
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CD25/IL-2R alpha Monoclonal specifically detects CD25/IL-2R alpha in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen CD25/IL-2R alpha
Applications Flow Cytometry
Classification Monoclonal
Clone PC61.5.3
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Alias CD25, CD25 antigen, IDDM10, IL-2 R alpha, IL-2 receptor subunit alpha, IL2R, IL2R alpha, IL-2R subunit alpha, IL-2Ra, IL2RA, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR
Gene Symbols IL2RA
Host Species Rat
Immunogen B6.1 mouse cytotoxic T cell line
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3559
Test Specificity Detects mouse CD25/IL-2 R alpha in flow cytometry.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.