Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CD27 Ligand/TNFSF7 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF783SP
This item is not returnable.
View return policy
Description
CD27 Ligand/TNFSF7/CD70 Polyclonal specifically detects CD27 Ligand/TNFSF7/CD70 in Mouse samples. It is validated for Western Blot, Neutralization.Specifications
CD27 Ligand/TNFSF7/CD70 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD27-L, CD27LCD27 ligand, CD27LG, CD70, CD70 molecule, Ki-24 antigen, surface antigen CD70, TNFSF7, TNFSF7CD70 antigen, tumor necrosis factor (ligand) superfamily, member 7, Tumor necrosis factor ligand superfamily member 7 | |
Goat | |
Affinity Purified | |
RUO | |
970 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Neutralization | |
LYOPH | |
Western Blot 0.1 ug/mL, Neutralization 3-12 ug/mL | |
O55237 | |
CD70 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse CD27 Ligand/TNFSF7 Gln47-Pro195 Accession # O55237 | |
25 μg | |
Primary | |
Detects mouse CD27 Ligand in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human CD27 Ligand is observed. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction