Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CD27/TNFRSF7 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB574SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB574SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB574SP Supplier R&D Systems Supplier No. MAB574SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CD27/TNFRSF7 Monoclonal specifically detects CD27/TNFRSF7 in Mouse samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen CD27/TNFRSF7
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 137910
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P41272
Gene Alias CD27, CD27 antigen, CD27 molecule, MGC20393, S152CD27L receptor, T cell activation antigen CD27, T cell activation antigen S152, T14, TNFRSF7, TNFRSF7T-cell activation antigen CD27, Tp55, Tumor necrosis factor receptor superfamily member 7, tumor necrosis factor receptor superfamily, member 7
Gene Symbols CD27
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CD27/TNFRSF7 Thr21-Arg182 Accession # P41272
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 939
Test Specificity Detects mouse CD27 in direct ELISAs and Western blots. In Western blots, does not cross-react with recombinant human (rh) CD27, rhCD30, rhCD40, rhDR3, rhDR6, rhFas Ligand, rhTRAIL R3, rhTRAIL R4, rhLT beta R, rhNGF-R1, rh4-1BB, rhGITR, rhTNFR1, rhTNF RII, rhBAFF, rhTAJ, rhEDA, rhOPG, rhRELT, rhRANK, recombinant mouse (rm) CD30, rmCD40, rmGITR, rmHVEM, rmRANK, rmFas, rmTRAIL R2, rm4-1BB, rmLT beta R, rmOPG, rmNGFR, rmDR3, rmBAFF R, rmEDAR, rmOX-40, rmTWEAK R, rmTNF R1, recombinant rat Fas, or recombinant feline Fas.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.