Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CD2F-10/SLAMF9 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF2829
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF2829 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF2829 Supplier R&D Systems Supplier No. BAF2829
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

CD2F-10/SLAMF9 Polyclonal specifically detects CD2F-10/SLAMF9 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen CD2F-10/SLAMF9
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. Q9D780
Gene Alias CD2 family member 10, CD2F10, CD2F-10MGC88194, CD84 homolog 1, CD84H1, CD84-H1, CD84-H1cluster of differentiation 2 antigen family member 10, cluster of differentiation 84 homolog 1, SF2001, signaling lymphocytic activation molecule family member 2001, signaling lymphocytic activation molecule family member 9, SLAM family member 9, SLAMF9
Gene Symbols SLAMF9
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CD2F-10/SLAMF9 Phe18-Leu230 Accession # Q9D780
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 89886
Test Specificity Detects mouse CD2F-10/SLAMF9 in Western blots. In Western blots, less than 5% cross-reactivity with recombinant mouse CD2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.