Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CD300a/LMIR1 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1186R100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1186R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1186R100 Supplier R&D Systems Supplier No. FAB1186R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CD300a/LMIR1 Monoclonal specifically detects CD300a/LMIR1 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen CD300a/LMIR1
Applications Flow Cytometry
Classification Monoclonal
Clone 172224
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. BAC80268
Gene Alias CD300a, CD300a antigenCMRF35H9, CD300a molecule, CLM8, CLM-8, CMRF-35H, CMRF35-H, CMRF35H leukocyte immunoglobulin-like receptor, CMRF35-H9, CMRF-35-H9CD300 antigen-like family member A, CMRF35HIRp60, CMRF35-like molecule 8, IgSF12, IGSF12NK inhibitory receptor, Immunoglobulin superfamily member 12, Inhibitory receptor protein 60, IRC1, IRC1/IRC2, IRC2, IRp60, leukocyte membrane antigen, LMIR1, MAIR-I, Mcipr1, NKRL, Pigr4
Gene Symbols CD300A
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CD300a/LMIR1 Leu28-Arg183 Accession # BAC80268
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11314
Test Specificity Detects mouse CD300a/LMIR1 in direct ELISAs. In direct ELISAs, approximately 2% cross-reactivity with recombinant mouse (rm) LMIR2 is observed and no cross-reactivity with recombinant human LMIR1, 2, 3, 4, 5, 6, or rmLMIR3, 4, or 5 is observed.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.