Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CD31/PECAM-1 Alexa Fluor 488-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody
Supplier: R&D Systems FAB6874G
This item is not returnable.
View return policy
Description
CD31/PECAM-1 Monoclonal specifically detects CD31/PECAM-1 in Mouse samples. It is validated for Flow Cytometry.Specifications
| CD31/PECAM-1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA', PECA1, PECAM1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5175 | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 693102 | |
| Flow Cytometry 5 uL/10^6 cells | |
| Q08481 | |
| PECAM1 | |
| NS0 mouse myeloma cell line transfected with mouse CD31/PECAM-1 Glu18-Lys590 Accession # Q08481 | |
| 100 Tests | |
| Primary | |
| Detects mouse CD31/PECAM-1 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) DCC, rmICAM-1, -2, -5, rmMAdCAM-1, rmVCAM-1, recombinant porcine CD31/PECAM-1, recombinant human (rh) CD31/PECAM-1, rhCEACAM-1, rhSIGIRR, rhICAM-3, or rhICAM-4 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction