Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CD34 Alexa Fluor 350-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB6518U100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB6518U100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB6518U100 Supplier R&D Systems Supplier No. FAB6518U100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CD34 Monoclonal specifically detects CD34 in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen CD34
Applications Flow Cytometry
Classification Monoclonal
Clone 700011
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 350
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q64314
Gene Alias CD34, CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule, HPCA1
Gene Symbols CD34
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse CD34 Thr35-Thr287 Accession # Q64314
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 947
Test Specificity Detects mouse CD34 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human, rat, or porcine CD34 is observed.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.