Learn More
Mouse CD34 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | CD34 |
| Applications | Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 488 |
| Dilution | Flow Cytometry 5 uL/10^6 cells |
| Gene Accession No. | Q64314 |
| Gene Alias | CD34, CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule, HPCA1 |
| Gene Symbols | CD34 |
| Host Species | Sheep |
| Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse CD34 Thr35-Thr287 Accession # Q64314 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.