Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse CD34 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB6518R100UG
This item is not returnable.
View return policy
Description
CD34 Monoclonal specifically detects CD34 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD34 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q64314 | |
CD34 | |
Mouse myeloma cell line NS0-derived recombinant mouse CD34 Thr35-Thr287 Accession # Q64314 | |
RUO | |
947 | |
Mouse | |
IgG2a |
Flow Cytometry | |
700011 | |
Alexa Fluor 647 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
CD34, CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule, HPCA1 | |
Rat | |
100 μg | |
Primary | |
Detects mouse CD34 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human, rat, or porcine CD34 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction