Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CD36 Mouse anti-Rat, Mouse, Clone: 63, MilliporeSigma™

Catalog No. MABF956MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF956MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF956MI Supplier MilliporeSigma Supplier No. MABF956
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect CD36 by using this mouse monoclonal Anti-Mouse CD36 Antibody, Clone 63. It is used in Flow Cytometry and Inhibition studies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse CD36
Applications Flow Cytometry, Inhibition Assays
Classification Monoclonal
Clone 63
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols cd36
Host Species Mouse
Immunogen Full-length adenovirus vector injected into CD36 null mice.
Purification Method Protein L
Quantity 200 μL
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_001153027
Test Specificity Clone 63 specifically detects CD36 in RAW264.7 cells.
Target Species Rat, Mouse
Content And Storage Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgA κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.